Learn More
Abnova™ Human ABCA10 Partial ORF (NP_525021, 1 a.a. - 80 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010349-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intracellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, and White). This encoded protein is a member of the ABC1 subfamily. Members of the ABC1 subfamily comprise the only major ABC subfamily found exclusively in multicellular eukaryotes. This gene is clustered among 4 other ABC1 family members on 17q24, but neither the substrate nor the function of this gene is known. [provided by RefSeq]
Sequence: MNKMALASFMKGRTVIGTPDEETMDIELPKKYHEMVGVIFSDTFSYRLKFNWGYRIPVIKEHSEYTEHCWAMHGEIFCYLSpecifications
NP_525021 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.54kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNKMALASFMKGRTVIGTPDEETMDIELPKKYHEMVGVIFSDTFSYRLKFNWGYRIPVIKEHSEYTEHCWAMHGEIFCYL | |
RUO | |
ABCA10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10349 | |
ABCA10 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EST698739 | |
ABCA10 | |
Recombinant | |
wheat germ expression system |