Learn More
Abnova™ Human A4GALT Partial ORF (NP_059132, 254 a.a. - 353 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00053947-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene catalyzes the transfer of galactose to lactosylceramide to form globotriaosylceramide, which has been identified as the P(k) antigen of the P blood group system. The encoded protein, which is a type II membrane protein found in the Golgi, is also required for the synthesis of the bacterial verotoxins receptor. [provided by RefSeq]
Sequence: LTRVFKKWCSIRSLAESRACRGVTTLPPEAFYPIPWQDWKKYFEDINPEELPRLLSATYAVHVWNKKSQGTRFEATSRALLAQLHARYCPTTHEAMKMYLSpecifications
NP_059132 | |
Liquid | |
53947 | |
A4GALT (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
A14GALT/A4GALT1/P1/PK | |
A4GALT | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LTRVFKKWCSIRSLAESRACRGVTTLPPEAFYPIPWQDWKKYFEDINPEELPRLLSATYAVHVWNKKSQGTRFEATSRALLAQLHARYCPTTHEAMKMYL | |
RUO | |
A4GALT | |
Wheat Germ (in vitro) | |
GST |