missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ HU 308

Product Code. p-4784026
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18023992 10 mg 10mg
18065212 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18023992 Supplier Tocris Bioscience™ Supplier No. 3088/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective CB2 agonist

Potent and selective CB2 receptor agonist (Ki values are 22.7 nM and > 10 μM for CB2 and CB1 receptors respectively, EC50 = 5.57 nM). Displays antiallodynic activity in the rat hindpaw incision model of postoperative pain. Also neuroprotective and improves motor performance in a mouse model of Huntington's Disease.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 256934-39-1
Molecular Formula C27H42O3

Specifications

Assay Percent Range >98%
Quantity 10 mg
Formula Weight 414.62
Molecular Weight (g/mol) 414.63
Chemical Name or Material HU 308
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.