Learn More
Tocris Bioscience™ HU 308
Product Code. 18023992
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18023992
Brand: Tocris Bioscience™ 3088/10
Potent and selective CB2 agonist
Potent and selective CB2 receptor agonist (Ki values are 22.7 nM and > 10 μM for CB2 and CB1 receptors respectively, EC50 = 5.57 nM). Displays antiallodynic activity in the rat hindpaw incision model of postoperative pain. Also neuroprotective and improves motor performance in a mouse model of Huntington's Disease.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 256934-39-1
Molecular Formula C27H42O3
Specifications
Assay Percent Range >98%
Quantity 10 mg
Formula Weight 414.62
Molecular Weight (g/mol) 414.63
Chemical Name or Material HU 308