Learn More
Trajan HT5 GC capillary column 0.22 mm ID x 0.1 µm film t
Brand:  Trajan 054638
HT5 GC capillary column 0.22 mm ID x 0.1 µm film thickness x 50 m length
Product Code. 18894864
  • 947.00€ / Each