Learn More
Applied Biosystems™ HT ExoSAP-IT™ Fast High-Throughput PCR Product Cleanup Reagent
Product Code. p-7092195
Click to view available options
No. of Reactions:
1000 rxns
23,040 rxn, 48 x 8-tube strips
480 rxns, 8-tube strip
5,760 rxn, 12 x 8-tube strips
5000 rxns
Unit Size:
1000 reactions
23040 reactions
480 reactions
5000 reactions
5760 reactions
Product Code. 15553687
Brand: Applied Biosystems™ 785951EA
Edge
Removes excess primers and dNTPs. Does not interfere with downstream applications. Ideal for high-throughput PCR product cleanup using liquid handling platforms and multi-channel pipettes.
  • Mixture of Exonuclease I and Shrimp Alkaline Phosphatase for the enzymatic removal of excess primers and dNTPs following a PCR reaction
  • One-tube PCR cleanup
  • Shortened 14-minute protocol
  • Multiple packaging formats (vials, 8-tube strips, 96-well plates) ensure ease of use and flexibility in experimental design
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at −20°C. Stable for one month at 4°C
No. of Reactions 480 rxns, 8-tube strip