Learn More
Proteintech HRP-Protein A
Shop All Proteintech Products
Click to view available options
Concentration:
0.2 mg/mL
Unit Size:
200µL
Description
This product is for research use only,not for diagnostic or therapeutic use
Specifications
Specifications
| Conjugate | HRP |
| Concentration | 0.2 mg/mL |
| Form | Liquid |
| Target | FC Fragment |
Product Content Correction
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion