Learn More
Description
The key component of the kit is a nitrocellulose membrane containing a Test line of immobilized Protein A and Protein G called a half strip. The HRP-antibody conjugate is run on the Protein A/G Strip. The conjugate binds the Protein A and Protein G concentrated on the Test line. After the addition of the HRP detection solution, a visible line on the strip appears.
Kit Contents:
- 1 cryovial of HRP detection solution
- 1 vial of Positive Control
- 3 cryovials of 10X Running Buffer
- 30 Protein A/G Strips
- Not supplied: 96-wells low binding plate, Bovine serum albumin (BSA)
Specifications
Specifications
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Quantity | 30 Tests |
| Target | HRP Check&Go! |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.