Learn More
Abnova™ HRAS (Human) Recombinant Protein
Human HRAS full-length ORF ( NP_005334, 1 a.a. - 189 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003265-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MTEYKLVVVGAGGVGKSALTIQLIQNHFVDEYDPTIEDSYRKQVVIDGETCLLDILDTAGQEEYSAMRDQYMRTGEGFLCVFAINNTKSFEDIHQYREQIKRVKDSDDVPMVLVGNKCDLAARTVESRQAQDLARSYGIPYIETSAKTRQGVEDAFYTLVREIRQHKLRKLNPPDESGPGCMSCKCVLS
Specifications
NP_005334 | |
v-Ha-ras Harvey rat sarcoma viral oncogene homolog | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HRAS | |
GST |
3265 | |
Wheat germ expression system | |
25 μg | |
C-BAS/HAS, C-H-RAS, C-HA-RAS1, CTLO, H-RASIDX, HAMSV, HRAS1, K-RAS, N-RAS, RASH1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |