Learn More
SCAT™ HPLC Supply and Waste Set, V2.0, Suitable for All HPLC Systems
Product Code. p-200026281 Shop All SCAT Europe Products
Click to view available options
Includes:
4 x Safety Cap III, GL45, 4 x air valve, 1 x Safety Waste Cap, S60/61, electrostatic conductive, 1 x Exhaust filter M, V3.0, with splash guard and change indicator, 1 x 5 liter canister, PE-HD-EL, 1 x Grounding cable, 2 x clamp, 1.5 m length, 4 x Laboratory bottle DURAN, GL45, 1000 mL, clear glass and round, with cap and pouring ring, 5 x PFA-fitting with integrated ferrule, 2.3 mm OD, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm OD, green, 10 x PFA fitting with integrated ferrule, 3.2 mm OD, blue, 10 x PFA blind plug, 2 x connector, straight, 6 to 8 mm ID, 1 x connector, curved, 5 to 11.5 mm ID, 1 x connector, angled, 9.5 to 10 mm ID
4 x Safety Cap III, GL45, 8 x air valve, 1 x Safety Waste Cap, GL45, 1 x Exhaust filter L, V3.0, with splash guard and change indicator, service life 12 months, 1 x 5 litre canister, PE-HD, 4 x Laboratory bottle DURAN, GL45, 1000 ml, clear glass and round, with cap and pouring ring , 5 x PFA-fitting with integrated ferrule, 2.3 mm OD, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm OD, green, 10 x PFA fitting with integrated ferrule, 3.2 mm OD, blue, 10 x PFA blind plug, 2 x connector, straight, 6 - 8 mm ID, 1 x connector, curved, 5.0 - 11.5mm ID, 1 x connector, angled, 9.5 - 10 mm ID
4 x Safety Cap III, GL45, 8 x air valve, 1 x Safety Waste Cap, S60/61, electrostatic conductive, 1 x Exhaust filter L, V3.0, with splash guard and change indicator, service life 12 month, 1 x 5 litre canister, PE-HD-EL, 1 x Grounding cable, 2x clamp, L = 1,5 m, 4 x Laboratory bottle DURAN, GL45, 1000 ml, clear glass and round, with cap and pouring ring , 5 x PFA-fitting with integrated ferrule, 2.3 mm OD, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm OD, green, 10 x PFA fitting with integrated ferrule, 3.2 mm OD, blue, 10 x PFA blind plug, 2 x connector, straight, 6 - 8 mm ID, 1 x connector, curved, 5.0 - 11.5mm ID, 1 x connector, angled, 9.5 - 10 mm ID
4 x Safety Cap III, V2.0, GL45, 1 x Safety Waste Cap, V2.0, GL45, 1 x Exhaust filter M, V2.0, 4 x Laboratory bottle DURAN, GL45, 1000 mL, 4 x air valve, 5 x PFA fitting with integrated ferrule, 2.3 mm, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm, green, 10 x PFA fitting with integrated ferrule, 3.2 mm, blue, 10 x Blanking plugs, 1.6/2.3/3.2 mm, colorless, 2 x Tube connector, straight 6 - 8 mm, 1 x Tube connector, curved, 5.0 - 11.5 mm, 1 x tube connector, angled, 9.5 - 10 mm
Unit Size:
1 set
1 sheet
Each
Product Code. 16458190
Brand: SCAT™ 307337
Only null left
Add to basket
Add to basket
The all-round carefree package suitable for all HPLC and UHPLC systems.
  • Protects your system from contamination
  • Keeps solvent mixtures stable
  • High performance materials with excellent chemical resistance
The safety set connects tubing safely and blocks hazardous vapours. Air valves and exhaust filters provide safe pressure equalisation without under- or overpressure in solvent bottles and waste containers
Especificaciones
Description Non-Electrostatic Conductive
For Use With (Equipment) Any HPLC System
Type HPLC System
Solvent Channels 8
Includes 4 x Safety Cap III, V2.0, GL45, 1 x Safety Waste Cap, V2.0, GL45, 1 x Exhaust filter M, V2.0, 4 x Laboratory bottle DURAN, GL45, 1000 mL, 4 x air valve, 5 x PFA fitting with integrated ferrule, 2.3 mm, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm, green, 10 x PFA fitting with integrated ferrule, 3.2 mm, blue, 10 x Blanking plugs, 1.6/2.3/3.2 mm, colorless, 2 x Tube connector, straight 6 - 8 mm, 1 x Tube connector, curved, 5.0 - 11.5 mm, 1 x tube connector, angled, 9.5 - 10 mm
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado