Learn More
SCAT HPLC Supply-Sets, V2.0
Product Code. 16443789 Shop All SCAT Europe Products
Click to view available options
Capacity (Metric):
100 mL
1 L
Color:
Brown
Clear
Material:
Glass (Round)
Glass (Square)
Glass (Youtility)
Solvent Channels:
1
2
3
Unit Size:
1 set
Product Code. 16443789
Brand: SCAT 307301
The HPLC Supply Sets, V2.0 of the manufacturer SCAT are combinations of equipment for contamination-free supply of HPLC systems with solvents. The sets consist of a laboratory bottle with GL 45 thread, a SCAT SafetyCap as well as capillaries, suction filters and air valves.
  • Customised system with high quality set components
  • SafetyCaps of the premium manufacturer SCAT
  • Laboratory bottles
  • Price attractive in comparison with single purchase
  • Ready for use, no further components necessary
SCAT offers a complete system for safe supply and disposal of solvents in analytics. All products are compatible across generations and perfectly matched to each other.
Specifications
Description HPLC Supply-Set I, V2.0: 1 x Safety Cap I GL45, 1x 1000 mL Laboratory bottle square & clear glass + 1 x 1,5 m capillary 3,2 mm OD, 1 x PTFE suction filter + air valve (317010)
For Use With (Equipment) Any HPLC System
Type Complete Solvent Collection System
Solvent Channels 1
Material Glass (Square)
Includes 1 x SafetyCap I GL45, 1x 1000 mL Laboratory bottle square and clear glass + 1 x 1.5 m capillary 3.2 mm OD, 1 x PTFE suction filter + air valve (317010)
Color Clear
Thread Size GL45
Capacity (Metric) 1 L