Learn More
Justrite™ HPLC Poly Manifold for Quick-Disconnect Disposal Cans
Product Code. 11997137
Click to view available options
Includes:
Manifold w/PP quick disconnect
Manifold w/SS quick disconnect
Manifold w/SS stacker connect
Material:
Polypropylene
Stainless Steel, Polypropylene
Unit Size:
Each
Product Code. 11997137
Brand: Justrite™ 28179
For use with Justrite™ Centura Quick-Disconnect HPLC Safety Disposal Cans
For use with Justrite™ Centura Quick-Disconnect HPLC Safety Disposal Cans
  • Enables up to six waste lines per manifold from HPLC processes to be fed into a single intake fitting
  • Six 1/4-28 ports and one 3/8 in. NPT port plus caps for unused ports
  • Tapped to accommodate 1/4-28 threads of separately available compression fittings
  • Multiple manifolds may be used on a custom-designed Quick Disconnect container as long as one disconnect remains unused to permit venting
  • Cat. No. 19-412-604 has a stacker connection enabling it to attach on top of Cat. No. 19-412-602 or 19-412-603; allows up to 12 lines to be fed into disposal can
  • Fittings sold separately (See 19-412-605, -606)
Specifications
Certifications/Compliance US OSHA, NFPA Code 30
Product Type Quick-disconnect Disposal Can
For Use With (Equipment) For Justrite™ centura quick-disconnect HPLC safety disposal cans
Material Stainless Steel, Polypropylene
Includes Manifold w/SS stacker connect
Quantity Ea.
Dimensions 7 x 5 x 4.5 in.
Length (English) 7 in.
Length (Metric) 17.8 cm
Width (English) 5 in.
Width (Metric) 12.7 cm
Height (English) 4.5 in.
Height (Metric) 11.4 cm