Learn More
Honeywell™ Howard Leight™ Leightning L1 Ear Muffs
Description
- Robust steel wire construction withstands demanding use, especially in tough environments
- Features patented Air Flow Control technology, delivering optimal attenuation across all frequencies
- Snap-in ear cushions make replacement quick and easy
- Multiple attenuation levels allow for targeted attenuation for a variety of applications
- Telescopic height adjustment remains fixed during use
- Delivers high degree of comfort; ideal for situations where compact earmuffs and reliable Noise-Blocking protection are required
Specifications
Specifications
| Color | Gray |
| Certifications/Compliance | ANSI S3.19-1974 |
| For Use With (Application) | Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, Metal fabrication, petrochemical Steel |
| Noise Reduction Rating (NRR) | 25 dB |
| Product Type | Ear Muffs |
| Dielectric | No |
| Style | Behind-the-neck |
| Includes | Elastic headband strap |