Learn More
Howard Leight™ Hygiene Kit for Ear Muffs
Click to view available options
:
Includes one pair replacement ear cussions and one pair foam inserts
Description
Includes
One pair of snap-in ear cushions and one pair of foam inserts
Recommended replacement every six months with general use, more frequently with heavy use.
Specifications
Specifications
| For Use With (Equipment) | All Howard Leight ear muffs and thunder T2s/T2H |
| Product Type | Hygiene Kit |
| Dielectric | Yes |
| Style | T3H |