Learn More
Howard Leight™ Hygiene Kit for Ear Muffs
Product Code. 12605023
Click to view available options
:
Includes one pair replacement ear cussions and one pair foam inserts
Product Code. 12605023
Brand: Honeywell Safety Products 1010975
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Includes
One pair of snap-in ear cushions and one pair of foam inserts
For extended performance and improved hygiene
Recommended replacement every six months with general use, more frequently with heavy use.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) All Howard Leight ear muffs and thunder T2s/T2H
Product Type Hygiene Kit
Dielectric Yes
Style T3H