Learn More
Honeywell™ Howard Leight™ Clarity™ C1 Earmuffs
Description
- Sound management technology
- Enhances on-the-job communication through uniform attenuation
- Noise Reduction Rating: 20dB
- Clarity blocks noise while voice, signal, alarm and other high frequencies can be heard more naturally, with less distortion, combating overprotection and worker isolation
- Dielectric/plastic construction withstands use and abuse, while protecting in electrical environments
- Quick-click height adjustment remains fixed during wear
- Snap-in ear cushions make replacement quick and easy
Specifications
Specifications
| Color | Metallic Blue |
| Certifications/Compliance | ISO 9001 / 2000 |
| For Use With (Application) | Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, Metal fabrication, petrochemical Steel |
| Noise Reduction Rating (NRR) | 20 dB |
| Product Type | Ear Muffs |
| Dielectric | Yes |
| Material | Plastic |
| Style | Over-the-head, folding |
| Foldable | Yes |