Learn More
Miele™ Housing unit, Aqua Purificator, stainless steel, for PG 8596 D AE DOS laboratory glass washers, Miele
Brand: Miele™ 10087340
This item is not returnable.
View return policy
Description
- Housing unit for process chemicals and dispenser modules
- Dimensions H 835mm (820), W 300mm, D 600mm
- Compatible with Miele glass laboratory washers models PG 8583, PG 8593
- Freestanding unit, can be built under if required
- Unit with removable door
- Outer panelling in stainless steel or white
- Interior dimensions H 690/380/285 mm (top drawer removed/lower/upper drawer), W 250mm, D 555mm /425mm (without/with drip tray and dispenser modules)
- Two levels: removable telescopic drawers with drip tray liners for canisters containing process chemicals
- Accommodates a total of 6 x 5L canisters (L 245mm x W 145mm x H 225 mm*).
- Lower drawer offers space for larger canisters: 2 x 10L (140 x 193 x 307mm, 223 x 203 x 321mm or 229 x 193 x 323mm) or 1 x 20L (289 x 233 x 396mm) or 1 x 25L (288 x 234 x 456mm)