Learn More
Thermo Scientific™ HotStart™ Storage Reaction Tubes
Product Code. 12318222 Shop All Thermo Scientific Products
Click to view available options
Product Line:
Hot Start Micro 50 Bulk
HotStart Micro 50 Strips
Volume (Metric):
0.2 mL
Unit Size:
Pack of 120
Pack of 480
Product Code. 12318222
Brand: Thermo Scientific™ 631011
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Eliminate tedious oil or wax overlays with the thin-walled Thermo Scientific™ HotStart™ Storage Reaction Tubes.
Features:
  • Bead is stable at room temperature
  • Assures synchronous reaction startup
  • PCR product can remain in tube for later analysis
  • Hinged caps provide snug seal
  • Presterilized; RNase- and DNase-free
Recommended for: Complex DNA or cDNA, very low copy number targets, more than 30 thermal cycles, multiple PCR or RT-PCR
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.2 mL
For Use With (Equipment) Thermal Cyclers
Product Line Hot Start Micro 50 Bulk
Sterility Sterile
Reaction Volume 25 to 50 μL
Product Type Tube