Learn More
Testo™ Hot Wire Probe
Click to view available options
Temperature Range (Metric):
-20°C to +70°C
Unit Size:
Each
Description
- Use the hot wire probe (thermal anemometer) to determine flow velocity and volume flow in ventilation ducts or at wall/ceiling outlets
- Ø 10 mm
- 4 functions in 1 probe: measurement of flow, temperature, humidity and absolute pressure
- With robust, non-rotatable telescope (can be extended to 1100 mm)
- The immersion depth is easy to read off thanks to the scaling on the telescope
- For easier handling, the probe can be bent up to an angle of 90°. On the handle there is a practical measuring button, which activates storage of the readings. So you can fully focus on the positioning of the probe in the ventilation duct
- Intelligent calibration concept
- The digital probe allows readings to be processed directly in the probe. This technology eliminates instrument measurement uncertainty
- Flow measuring range: 0 to +20 m/s
Specifications
Specifications
| Accuracy | Temperature - ±0.5 °C; Humidity - ±(1.8 %RH + 0.7 % of mv); Pressure - ±3 hPa, Velocity - ±(0.03 m/s + 4 % of mv) |
| Humidity Range | Humidity: 0 to 100%, Pressure - +700 to +1100 hPa; Velocity - 0 to +20 m/s |
| Temperature Range (Metric) | -20°C to +70°C |
| Length (Metric) | 1100 mm |