Learn More
Equip Labo™ Hot/Cold Water Gooseneck Tap
Product Code. 12648832
Change view
Click to view available options
Mounting:
Backsplash
Table
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mounting Unit Size
12648832 Backsplash Each
12611872 Table Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12648832 Supplier Equip Labo™ Supplier No. 4759
  • ONLINE EXCLUSIVE PRICE
    241.00€
    201.21€ / Pack of 1
    Save 39.79€
    16% Off
 Restricted Item
Estimated Shipment: 20-11-2026
With a height of 250mm, this cold/hot water tap is a common accessory for laundry and wet benches.
  • Common accessory for laundry and wet bench
  • Hot / cold gooseneck mixer on backsplash
  • Height 250mm
Specifications
Mounting Backsplash
Quantity 1 Ea.