Learn More
Hirschmann™ Hose Adapter
Product Code. p-7162678
Click to view available options
Color:
Gray
Green
Unit Size:
Each
Product Code. 10010461
Brand: Hirschmann™ 9903106
Only null left
Add to basket
Add to basket
Spare part for pipetus™ standard, hose connectors to pump
  • Spare part for pipetus™ standard, hose connectors to pump
Specifications
Color Gray
For Use With (Equipment) Pipetus Standard