Learn More
MP Biomedicals™ Hormones: Hydrocortisone
Product Code. 11454942 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5mg
1 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454942 5mg 5g
11464942 10 g 10g
11474942 25 g 25g
11444942 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11454942 Supplier MP Biomedicals™ Supplier No. 0219456905
 Restricted Item
Estimated Shipment: 24-11-2026
For research use only. Not intended for therapeutic use.
Specifications
Content And Storage 15° to 30°C
Product Type Hydrocortisone
Sterility Sterile
Quantity 5mg
Source Soy beans
compliance-icons
Product Identifier
  • Hydrocortisone, Cell Culture Reagent
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Hydrocortisone