Learn More
Fisherbrand™ Horizontal Large Gel Systems: Combs
Product Code. 11958726 Shop All Thermo Scientific Products
Click to view available options
Teeth Count:
1
8
12
16
20
24
28
32
36
Thickness (Metric) Teeth:
0.1 cm
0.15 cm
20 cm
Unit Size:
Each
Product Code. 11958726
Brand: Fisherbrand™ A232C
For large gel systems
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl A2 and A5 Large Gel Electrophoresis System
Type Standard Comb
Gel Dimensions 25 x 20 cm
Teeth Count 32
Thickness (Metric) Teeth 0.1 cm
Volume (Fill) Metric 20 μL
Length (English) Gel 9.8 in.
Length (Metric) Gel 25 cm
Width (English) Gel 7.8 in.
Width (English) Teeth 0.15 in.
Width (Metric) Gel 20 cm
Width (Metric) Teeth 0.4 cm