Learn More
Fisherbrand™ Horizontal Large Gel Systems: Combs
Shop All Thermo Scientific Products
Click to view available options
Teeth Count:
1
8
12
16
20
24
28
32
36
Thickness (Metric) Teeth:
0.1 cm
0.15 cm
20 cm
Unit Size:
Each
Description
For large gel systems
Specifications
Specifications
| For Use With (Equipment) | Owl A2 and A5 Large Gel Electrophoresis System |
| Type | Standard Comb |
| Gel Dimensions | 25 x 20 cm |
| Teeth Count | 16 |
| Thickness (Metric) Teeth | 0.15 cm |
| Volume (Fill) Metric | 75 μL |
| Length (English) Gel | 9.8 in. |
| Length (Metric) Gel | 25 cm |
| Width (English) Gel | 7.8 in. |
| Width (English) Teeth | 0.41 in. |
| Show More |