Learn More
Nabertherm™ Horizontal High-Temperature Tube Furnace
Product Code. 17398109 Shop All Nabertherm Products
Change view
Click to view available options
Diameter (Metric) Process Tube:
50 mm
80 mm
120 mm
Temperature (Metric):
1600°C
1700°C
1800°C
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Process Tube Temperature (Metric) Unit Size
17398109 80 mm 1700°C Each
17308119 80 mm 1800°C Each
17358109 50 mm 1600°C Each
17368109 50 mm 1700°C Each
17378109 50 mm 1800°C Each
17388109 80 mm 1600°C Each
17318119 120 mm 1600°C Each
17328119 120 mm 1700°C Each
17338119 120 mm 1800°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 17398109 Supplier Nabertherm™ Supplier No. RH281V7AN
 Restricted Item
Estimated Shipment: 17-02-2027
High-temperature horizontal tube furnaces. High-quality insulation materials made of vacuum-formed fiber plates enable energy-saving operation and a fast heating time due to low heat storage and heat conductivity. Different gas supply systems available.
  • Use: horizontal
  • Electrical connection: 3-phase
Please Note: A furnace below 2.9 kW will be delivered with a standard UK plug. Anything above 2.9 kW will be delivered with the appropriate CEE plug. For anything above 2.9 kW, please ensure you have the required socket prior to purchase.
Specifications
Controller Type P570
Description Horizontal
Heated Zone 530 mm
Temperature (Metric) 1700°C
Dimensions (L x W x H) Exterior 550 x 620 x 640 mm
Diameter (Metric) Process Tube 80 mm
Weight (Metric) 90 kg