Learn More
Honeywell™ Optifit Full Face Mask
Product Code. 11896950
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11896950 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11896950 Supplier Honeywell™ Supplier No. 1715011
  • ONLINE EXCLUSIVE PRICE
    281.40€
    135.27€ / Pack of 1
    Save 146.13€
    51% Off
View all promotions
 Restricted Item
Estimated Shipment: 23-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Effective respiratory protection in a full-face mask. Honeywell™ Optifit Full Face Mask offers dedicated respiratory protection depending on the canister type assembled to it.
  • Reusable APR with respiratory protection dictated by canister type selected.
  • For optimal protection, Sperian™ Aluminum Canisters or Sperian™ Plastic Canisters are recommended.
  • Note: Can only be used in sufficiently ventilated premises where the atmoshpere contains a minimum oxygen volume of 17%.
  • U-shaped silicone skirt.
  • Full lens with optimal lateral and downward vision.
  • Free choice of filter
  • Easy care
TRUSTED_SUSTAINABILITY
Specifications
Cartridge Type Filter
Certifications/Compliance EC Category PPE: 3, Quality Assurance: ISO 9001/2000, EC Certification Number: 0070-003-079-01-02-0009 - Ext. 01-10-03, EC Attestation Number 0070-003-079-01-02-0009 - Ext. 01-10-03, Declaration of ocnformity: INRS-1715011, EN 136
Product Type Optifit Full Face Mask
Size Medium
Facepiece Type Optifit
For Use With (Application) For industry uses
Includes Silicone Skirt/Dual Filter/ Diaphragm
Quantity 1 Ea.