Learn More
Honeywell™ Mont-Blanc S3 Safety Shoes
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
Unit Size:
Each
Pack of 2
Specifications
Specifications
| Certifications/Compliance | European Directive 89.686 CEE, EN 345-1, ISO 9001:2008 |
| Color | White |
| For Use With (Application) | For food services, pharmaceutical and medical use |
| Material | Leather |
| Size | 45 |
| Sole Features | Anti-puncture Steel midsole, white color, made up of double density Polyurethane |
| Sole Material | Double density Polyurethane |
| Style | Slip-on safety |
| Toe Material | Metallic |
| Resistant To | Impact, splash, slip-resistant |
| Show More |