Learn More
Sperian™ Honeywell™ Mont Blanc Safety Shoes
Product Code. 12671293
Click to view available options
Size:
35
36
39
41
42
44
45
46
47
Unit Size:
Each
Product Code. 12671293
Brand: Sperian™ 623321635/7
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Moccasin-style slip-on leather shoe with limited stitching on the upper. Sperian™ Honeywell™ Mont Blanc Safety Shoes have a double density injected outsole for extra protection.
  • Lined and padded collar
  • Anti-puncture midsole
  • Metallic toe cap 200J
  • Perforated insole, polyurethane white
  • Protects against impacts, slips and splashes
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance European Directive 89.686 CEE, EN 345-1, ISO 9001:2008
Color White
For Use With (Application) For food services, pharmaceutical and medical use
Material Leather
Size 35
Sole Features Anti-puncture Steel midsole, white color, made up of double density Polyurethane
Sole Material Double density Polyurethane
Style Slip-on safety
Toe Material Metallic
Resistant To Impact, splash, slip-resistant
Product Type Safety Shoe