Learn More
Sperian™ Honeywell™ QB1HYG Inner-Aural Banded Earplugs
Product Code. 11882701
Click to view available options
:
Color: Orange and Yellow
Product Code. 11882701
Brand: Sperian™ 3301282
Only null left
Add to basket
Add to basket
Smooth, ergonomic ear pods fit in the ear canal for maximum banded protection. Sperian™ Honeywell™ QB1HYG Inner-Aural Banded Earplugs are designed for work environments with intermittent noise.
  • Lightweight and portable
  • Reusable when washed with warm water and allowed to air dry
  • Patended band design prevents ear pods from touching dirty or contaminated surfaces when set down
TRUSTED_SUSTAINABILITY
Specifications
Color Orange, Yellow
Size Universal
Quantity 10/Pk.
Plug Shape Bell
Product Type Ear Plugs