missing translation for 'onlineSavingsMsg'
Learn More

Homovanillic Acid Sulfate Sodium Salt-d3, TRC

Product Code. 30407658
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407658 10 mg
30408362 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30407658 Supplier Toronto Research Chemicals Supplier No. TRCH66950710MG
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C9H5D3Na2O7S
Molecular Weight (g/mol) 309.22
Synonym 3-(Methoxy-d3)-4-(sulfooxy)-benzeneacetic Acid Sodium Salt
SMILES O=C(O[Na])CC1=CC(OC([2H])([2H])[2H])=C(OS(=O)(O[Na])=O)C=C1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H10O7S.2Na/c1-15-8-4-6(5-9(10)11)2-3-7(8)16-17(12,13)14;;/h2-4H,5H2,1H3,(H,10,11)(H,12,13,14);;/q;2*+1/p-2/i1D3;;
Chemical Name or Material Homovanillic Acid Sulfate Sodium Salt-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.