Learn More
Schuett Biotec™ Homgen™ Semi-Automatic Homogenizer
Product Code. 10190771
Change view
Click to view available options
Voltage:
230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
10190771 230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10190771 Supplier Schuett Biotec™ Supplier No. 3201011
  • ONLINE EXCLUSIVE PRICE
    7371.00€
    6397.36€ / Pack of 1
    Save 973.64€
    13% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Specifications
Voltage 230 V
Includes Cooling vessel, universal quick-grip chuck and univversal clamping adapter