Learn More
Karl Hecht Wooden blockwith3 brown pipette bottles 50 g.
Brand:  Karl Hecht 42611020
Wooden blockwith3 brown pipette bottles 50 g.
Product Code. 10023871
  • ONLINE EXCLUSIVE PRICE
    88.25€
    72.04€ / Each
    Save 16.21€
    18% Off
Only null left
Estimated Shipment: 29-01-2026