Learn More
Hoefer™ TE70X and TE77X Semidry Blotting Systems
Product Code. 11915348
Change view
Click to view available options
Type:
Semi Dry Electroblotting System
Includes:
21 x 26 cm Blot Paper
Built-in power supply, Molded base with Platinum coated Titanium anode and high voltage Leads, hinged lid with Stainless Steel cathode, power cables, blotter Paper (21 x 26cm) 25 sheets, porous cellophane (20 x 35.5cm) 50 sheets, two solid Mylar™
Power Supply, 14 x 16 cm Blot Paper
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Includes Unit Size
11915348 Semi Dry Electroblotting System Built-in power supply, Molded base with Platinum coated Titanium anode and high voltage Leads, hinged lid with Stainless Steel cathode, power cables, blotter Paper (21 x 26cm) 25 sheets, porous cellophane (20 x 35.5cm) 50 sheets, two solid Mylar™ Each
11905348 Semi Dry Electroblotting System Power Supply, 14 x 16 cm Blot Paper Each
11995338 Semi Dry Electroblotting System 21 x 26 cm Blot Paper Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11915348 Supplier Hoefer™ Supplier No. TE77XP
  • ONLINE EXCLUSIVE PRICE
    2865.00€
    2579.45€ / Pack of 1
    Save 285.55€
    9% Off
View all promotions
Estimated Shipment: 01-02-2027
Includes
Molded base with iridium oxide anode, top assembly with stainless-steel cathode, power cables, 25 sheets of blotting paper, 50 sheets of cellophane (Mfr. Nos. TE70X and TE70XP only), two solid Mylar™ masks
Perform uniform transfers of proteins and nucleic acids from polyacrylamide and agarose gels in less than one hour
  • Transfer up to two polyacrylamide gels in a stack
  • Use minimal buffer
  • Do not generate the excessive heat that can distort bands or denature protein
  • Utilize durable iridium oxide and stainless-steel electrodes to assure uniform, contamination-free transfers
  • Offer automatic transfer-stopping feature to prevent overheating when buffer is depleted
  • Available with built-in power supply to save space and simplify transfer
Certifications
CE marked, CSA C22.2 1010.1, EN61010-1, UL3101-1
Specifications
Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Content And Storage Built-in power supply, Molded base with Platinum coated Titanium anode and high voltage Leads, hinged lid with Stainless Steel cathode, power cables, blotter Paper (21 x 26cm) 25 sheets, porous cellophane (20 x 35.5cm) 50 sheets, two solid Mylar™
For Use With (Equipment) Electrophoresis Blotting System
Type Semi Dry Electroblotting System
Humidity Range Up to 80%
Includes Built-in power supply, Molded base with Platinum coated Titanium anode and high voltage Leads, hinged lid with Stainless Steel cathode, power cables, blotter Paper (21 x 26cm) 25 sheets, porous cellophane (20 x 35.5cm) 50 sheets, two solid Mylar™
Quantity Power Supply, 21 x 26 cm Blot Paper
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Dimensions (L x W x H) 18.89 x 15 x 4 in. (48 x 38 x 10 cm)
Length (English) 18.89 in.
Length (Metric) 48 cm
Width (English) 15 in.
Width (Metric) 38 cm
Height (English) 4 in.
Height (Metric) 10 cm
Voltage 30 V
Wattage 15 w
Amperage 500 mA