Learn More
Hoefer™ TE22 Mini Tank Blotting Unit
Product Code. 10380895
Change view
Click to view available options
Type:
Mighty Small™ Transfer Tank
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10380895 Mighty Small™ Transfer Tank Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10380895 Supplier Hoefer™ Supplier No. TE22
  • ONLINE EXCLUSIVE PRICE
    1270.00€
    1235.61€ / Pack of 1
    Save 34.39€
    2% Off
View all promotions
Estimated Shipment: 10-11-2026
Includes
Buffer chamber, lid with leads, two electrode panels, four gel cassettes, eight 0.13 in. thick foam sponges, four 0.25 in. thick foam sponges, 25 sheets blotting paper
Safe, economical unit requires minimal current and only about 1L of buffer
  • Built-in alumina heat exchanger permits efficient transfers in less than one hour
  • Platinum electrode ensures uniform electrical field for complete and even transfers
  • Hinged, color-coded cassettes enable simple loading and easy orientation of gel sandwich
  • Alumina-covered serpentine channel in base provides excellent temperature control with no more than 5°C rise during a normal run
Certifications
CE marked, CSA C22.2 1010.1, EN61010-1, UL3101-1
Specifications
Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Device Holds Up to 4, 9 x 10 cm Gel
Type Mighty Small™ Transfer Tank
Humidity Range Up to 80%
Includes Lower Buffer chamber with Alumina heat exchanger, safety lid with high voltage Leads, two electrode panels, four gel cassettes, eight 0.13 in. thick Foam sponges, four 0.25 in. thick Foam sponges, 25 sheets blotting Paper, cassette hook
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Volume (English) Buffer ≈33.81 oz.
Volume (Metric) Buffer ≈1000 mL
Dimensions (L x W x H) 6.49 x 5.51 x 9.44 in. (16.5 x 14 x 24 cm)
Length (English) 6.49 in.
Length (Metric) 16.5 cm
Width (English) 5.51 in.
Width (Metric) 14 cm
Height (English) 9.44 in.
Height (Metric) 24 cm
Voltage 100 V
Wattage 200 w
Amperage 500 mA