Learn More
Hoefer™ SUB15 Standard Submarine Running Tray
Description
- 15 x 15 cm
- Casting Gates
Specifications
Specifications
| For Use With (Equipment) | SUB15 Standard Submarine |
| Type | Running Tray |
| Includes | Casting gate |
| Dimensions (L x W) | 5.9 x 1.96 in. (15 x 5 cm) |
| Length (English) | 5.9 in. |
| Length (Metric) | 15 cm |
| Width (English) | 1.96 in. |
| Width (Metric) | 5 cm |