Learn More
Hoefer™ SUB10 Mini-Plus Submarine Running Tray
Product Code. 12695726
Change view
Click to view available options
For Use With (Equipment):
SUB10 Mini plus Submarine Unit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12695726 SUB10 Mini plus Submarine Unit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12695726 Supplier Hoefer™ Supplier No. SUB10UT
Only null left
Add to basket
Add to basket
Accessories for SUB10 Mini-Plus Submarine
  • 10 x 11.5 cm
  • Silcone Gasket
Specifications
For Use With (Equipment) SUB10 Mini plus Submarine Unit
Type Running Tray
Includes Silicone gasket
Dimensions (L x W) 4.52 x 3.93 in. (11.5 x 10 cm)
Length (English) 4.52 in.
Length (Metric) 11.5 cm
Width (English) 3.93 in.
Width (Metric) 10 cm