Learn More
Hoefer™ SE 900 Large Format Vertical Electrophoresis System
Product Code. 11995368
Click to view available options
Includes:
Tank, Tubing, Lid, PAGE Rack, Caster, Cassettes
Unit Size:
Each
Product Code. 11995368
Brand: Hoefer™ SE9001.0
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Includes
  • Buffer chamber with pumps, safety lid with leads, gel rack, 10 ft. tubing, two pair recirculation bath connectors, drain connector, seven tubing clamps, gel seal, wonder wedge plate separation tool
  • Mfr. No. SE900-1.0 also includes: Multicaster with ported face plate, six red spring clamps, two thumb screws, ten divider sheets, filler plug, cassette blank, gel seal, six glass hinge casettes
Ideal for use with 24cm IPG strips in the second dimension of 2-D electrophoresis
  • Runs 1 to 6 gels
  • Uses hinged cassettes for convenience and straight dye fronts
  • Easy to use — no clamps, no loose parts, bottom drain port
  • Utilizes internal pump for even temperature distribution
  • Compatible with a recirculating chiller for active cooling
Certifications
UL61010-1, CSAC22.2 No. 61010-1, EN61010-1:2001, CE Marked
Specifications
Certifications/Compliance EN61010-1, UL61010-1, CSA C22.2, CE
Type Vertical Electrophoresis System
Humidity Range Up to 80%
Includes Tank, Tubing, Lid, PAGE Rack, Caster, Cassettes
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Temperature (Metric) Storage 15°C to 30°C
Width (English) 16.92 in.
Width (English) Gel 9.84 in.
Width (Metric) 43 cm
Width (Metric) Gel 25 cm
Height (English) 16.92 in.
Height (English) Gel 7.87 in.
Height (Metric) 43 cm
Depth (English) 7.87 in.
Depth (Metric) 20 cm
Weight (English) 29.98 lb.
Weight (Metric) 13.6 kg
Voltage 600 V
Wattage 150 w
Amperage 1000 mA