Learn More
Hoefer™ Replacement Parts for HE 99X Max Submarine Electrophoresis Unit
Click to view available options
Type:
Buffer Chamber Assembly
Lid Assembly
Unit Size:
Each
Description
Replacement Parts
- Buffer chamber assembly (Includes 4 leveling feet, 2 banana plugs with nickel and nylon washers, and 2 platinum wire electrode strip assemblies)
- Lid assembly (Includes lid, red and black high-voltage leads, and two lid plugs)
Specifications
Specifications
| For Use With (Equipment) | HE99X Max Submarine Electrophoresis Unit |
| Type | Buffer Chamber Assembly |
| Includes | 4 Leveling feet, 2 banana plugs with Nickel and Nylon washers and 2 Platinum wire electrode strip assemblies |