Learn More
Hoefer™ miniVE Electrophoresis Unit Accessories and Parts
missing translation for 'orderingAttributeHoverText'
Type:
Banana Plug Kit
Clamp Assembly
Clamp Screw
Lid Assembly
Lower Buffer Chamber
Sealing Plate Assembly
Upper Buffer Chamber Assembly
missing translation for 'unitSize'
Each
Pack of 5
Description
- Accesories include High Voltage Leads (2 pcs), Banana Plug Kit (2 plugs, 2 nuts, 4 washers), Foam Gasket, Clamp Screws (5 pcs), Upper Buffer Chamber Assembly, Clamp Assembly, Sealing Plate Assembly, Sealing Gasket and Lower Buffer Chamber (tank)
Specifications
Specifications
| For Use With (Equipment) | MiniVE Vertical Electrophoresis System |
| Type | Sealing Plate Assembly |
| Includes | Sealing gasket |