Learn More
Hoefer™ miniVE Electrophoresis Unit Accessories and Parts
Part and accessories for miniVE Electrophoresis Unit
Brand:  Hoefer™ SE313
Product Code. 11559703
  • 30.20€ / Pack of 1
Only null left
Estimated Shipment: 17-06-2026
Description
  • Accesories include High Voltage Leads (2 pcs), Banana Plug Kit (2 plugs, 2 nuts, 4 washers), Foam Gasket, Clamp Screws (5 pcs), Upper Buffer Chamber Assembly, Clamp Assembly, Sealing Plate Assembly, Sealing Gasket and Lower Buffer Chamber (tank)
Specifications
Specifications
MiniVE™ Vertical Electrophoresis System
Sealing Gasket