Learn More
Hoefer™ Heat Exchanger (Lower Electrode Assembly) for SE 600 Series Electrophoresis Unit
Description
Includes
Banana plug and Platinum wire anode
- For use with SE 600 Series Electrophoresis Unit
Specifications
Specifications
| For Use With (Equipment) | SE 600 Series Electrophoresis Unit |
| Type | Heat Exchanger (Lower Electrode Assembly) |
| Includes | Banana plug and Platinum wire anode |