Learn More
Hoefer™ Glass Plate
Product Code. 11574404
Change view
Click to view available options
Quantity:
2 Each
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11574404 2 Each Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11574404 Supplier Hoefer Supplier No. SE6402LF
Only null left
Add to basket
Add to basket
They are lightweight and virtually unbreakable when used with reasonable care
All combs and spacers are moulded or fabricated of durable plastics highly resistant to acids, bases and all common buffer systems.
They are lightweight and virtually unbreakable when used with reasonable care.Preparative combs have adjustable comb back, required to convert any 25mm depth comb to 10mm or 15mm depth.
Specifications
Type Glass Plate
Gel Dimensions 18 x 8 cm
Quantity 2 Each