Learn More
Hoefer™ GD 2000 Vacuum Gel Dryer Accessories: Rubber Overlay Sheet
Product Code. 10848910 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
GD 2000 Vacuum Gel Dryer System, SE 1140, SE 1150, SE 1160 and SE 1175 Gel Dryers
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10848910 GD 2000 Vacuum Gel Dryer System, SE 1140, SE 1150, SE 1160 and SE 1175 Gel Dryers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10848910 Supplier Hoefer™ Supplier No. SE1143
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Dries acrylamide and agarose gels rapidly, evenly and safely.
  • Clear Silicone Rubber Overlay Sheet
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
For Use With (Equipment) GD 2000 Vacuum Gel Dryer System, SE 1140, SE 1150, SE 1160 and SE 1175 Gel Dryers
Type Rubber Overlay Sheet
Material Silicone