Learn More
Hoefer™ Combs for SUB25 Maxi-Plus Submarine Gel Electrophoresis Unit
Description
- Either 26 or 52 Wells
- 1, 1.5 and 2 mm Thickness
- 3 or 7 mm Well Width
- Min. 13, max. 60 μL Sample Volume
Specifications
Specifications
| For Use With (Application) | 52 sample throughput (multi-channel pipette compatible) |
| For Use With (Equipment) | SUB25 Maxi-Plus Unit |
| Thickness (English) | 0.07 in. |
| Thickness (Metric) | 2 mm |
| Volume (English) | 0.0008 oz. |
| Volume (Metric) | 25 μL |
| Width (English) Gel | 9.84 in. |