Learn More
Cytiva HiTrap™ Capto™ Q ImpRes IEX Columns
Shop All Cytiva Products
Click to view available options
Quantity:
5 x 1 mL
5 x 5 mL
Flow Rate:
<20 mL/min.
<4 mL/min.
Unit Size:
Pack of 5
Description
- Combined properties of high binding capacity, high flow rate and low back pressure reduces process cycle times and improves productivity at process scale
- Rigid agarose matrix enables high bed heights and purification of viscous samples at high flow rates
- Screening of selectivity, binding and elution conditions, as well as small scale purifications
- Strong quartenary amine anion exchanger
Specifications
Specifications
| Description | HiTrap Capto Q ImpRes, 5 x 5 mL |
| Flow Rate | <20 mL/min. |
| For Use With (Application) | HiTrap Capto Q ImpRes chromatography column is packed with a strong anion exchange modern resin for high resolution, small-scale protein purification. |
| Content And Storage | 4°C to 30°C in 20% Ethanol |
| Diameter (Metric) Inner Column | 7 mm |
| Volume (Metric) | 5 mL |
| Quantity | 5 x 5 mL |
Safety and Handling
Signal Word
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.