Learn More
Epredia™ HistoStar™ Heated Forceps
Product Code. 12638216 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Heated Forceps, 1 mm tip
Heated Forceps, 2 mm tip
Heated Forceps, 4 mm tip
Color:
Blue
Red
Yellow
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Color Unit Size
12638216 Heated Forceps, 4 mm tip Blue Each
12618216 Heated Forceps, 1 mm tip Red Each
12628216 Heated Forceps, 2 mm tip Yellow Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12638216 Supplier Epredia™ Supplier No. A81010035
  • ONLINE EXCLUSIVE PRICE
    687.00€
    535.70€ / Pack of 1
    Save 151.30€
    22% Off
Estimated Shipment: 04-02-2027
Epredia™ Heated Forceps work in conjunction with the Epredia™ HistoStar™ Embedding Workstation. Multiple sizes are available.
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
For Use With (Equipment) Epredia™ HistoStar™ Embedding Workstation
Product Type Heated Forceps, 4 mm tip