Learn More
Epredia™ HistoStar™ Embedding Workstation Accessories
Shop All Epredia Products
Click to view available options
Product Type:
Attachable Magnifier Assembly
Foot Switch
Spatula Kit
Tamping Tool Kit
Unit Size:
Each
Pack of 2
Description
Today's embedding centers can be uncomfortable, hot, or poorly illuminated, leading to fatigue and potential errors. The new Epredia HistoStar Embedding Workstation allows users to embed comfortably and efficiently. With brightly illuminated work surfaces, cool and contoured arm rests and high throughput capacity, this workstation delivers significant improvements in ergonomics and efficiency.
Brightly illuminated and clutter free
- LED lighting uniformly illuminates the workspace without the clutter of awkward remote lamps
- A properly illuminated workspace reduces fatigue and minimizes errors
- Lighting intensity is easily user adjustable
- All user contact points are smooth and insulated, providing a cool and comfortable workspace
- Pressure points and uncomfortable heat are eliminated
- The capacity of the HistoStar can keep up with even the busiest laboratories
- Capacity will never limit your workflow with 5 liter paraffin capacity, coldplate area for 72 base molds, large heated workspace and specimen holding area
Any sample size handled with ease
- The HistoStar offers the first adjustable paraffin dispense paddle
- A simple adjustment moves the paddle to the most ergonomic position for the user
- Even Super Mega Cassettes can be embedded with ease
- The HistoStar is the only workstation with a heated wax trimmer built directly into the workspace
- Excess paraffin can be removed at the embedding station, eliminating the need for additional equipment or wax scrapping at the microtome
- Large, easy-to-read touchscreen display allows quick access to the temperature controls and other parameters
- Programmable asleep mode saves energy around your workflow
- The robust HistoStar design minimizes moving parts, reducing wear and service requirements
- Ensures dependable performance for years to come
Specifications
Specifications
| For Use With (Equipment) | HistoStar Embedding Center |
| Product Type | Foot Switch |