Learn More
Epredia™ HistoScreen™ Cassettes in Tube Packs
Product Code. 16393936 Shop All Epredia Products
Click to view available options
Color:
Beige
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
missing translation for 'unitSize'
Pack of 750
Product Code. 16393936
missing translation for 'mfr': Epredia™ B851001LG
Eliminates the need for sponges or bags, ideal for very small biopsy tissue samples. Cassettes are compatible with Epredia PrintMate™ Cassette Printers, have a back-edge lid hinge, and loose-packed lids that are available in 12 color varieties.
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Mesh
Closure Included Yes
Quantity 750/Cs.
Closure Type Separate Lid, Back Hinge
PrintMate Compatible Yes
Color Light Green
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type HistoScreen Cassette