Learn More
Epredia™ HistoScreen™ Cassettes
For very small biopsy tissue samples with a back-edge lid hinge. Eliminates the need for sponges or biopsy bags.
660.00€
Specifications
Closure Included | Separate lids included |
---|---|
Pore Style | Mesh |
Closure Type | Back Hinge |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
16333926
|
Epredia™
B851000BL |
Blue |
660.00€
Pack of 1000 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
16323926
|
Epredia™
B851000BE |
Beige |
660.00€
Pack of 1000 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
16353926
|
Epredia™
B851000GY |
Gray |
660.00€
Pack of 1000 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
16343926
|
Epredia™
B851000GN |
Green |
660.00€
Pack of 1000 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
16363926
|
Epredia™
B851000LA |
Lavender |
660.00€
Pack of 1000 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
16373926
|
Epredia™
B851000LG |
Light Green |
660.00€
Pack of 1000 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
16383926
|
Epredia™
B851000LI |
Lilac |
660.00€
Pack of 1000 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
16393926
|
Epredia™
B851000OR |
Orange |
660.00€
Pack of 1000 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
16303936
|
Epredia™
B851000PK |
Pink |
660.00€
Pack of 1000 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
16313936
|
Epredia™
B851000TN |
Tan |
660.00€
Pack of 1000 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
16323936
|
Epredia™
B851000WH |
White |
660.00€
Pack of 1000 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
16333936
|
Epredia™
B851000YW |
Yellow |
660.00€
Pack of 1000 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
- Surface finish and reinforced print faces are engineered to optimize print quality. Tube design is precisely engineered to reduce jamming and prevent downtime.
- Grossing, Processing and Embedding – cassette design and flow characteristics tested to ensure efficient reagent exchange. Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples.
- Precision engineering for solid connection to microtomes.
- Cassette surface texture optimizes scanability for archiving.
Specifications
Separate lids included | |
Mesh | |
Back Hinge |