Learn More
Heinz Herenz HistoPlast Paraffin Wax
Product Code. 30158402
Click to view available options
Melting Point:
54°C to 56°C.
56°C to 58°C
58°C to 60°C
60°C to 62°C
Unit Size:
Pack of 10
Product Code. 30158402
Brand: Heinz Herenz 1131237
HistoPlast paraffin wax, supplied in pearl shape. For tissue section infiltration and embedding.
  • HistoPlast paraffin wax
  • Supplied in pearl shape
  • For tissue section infiltration and embedding
  • Compound of purified paraffin and plastic polymers, without dimethyl sulfoxide (DMSO).
  • Cutting thickness 2μm.
Specifications
Melting Point 60°C to 62°C
Product Type Paraffin
Form Pellet
compliance-icons compliance-icons
Product Identifier
  • X10 HistoPlast Paraffin Wax, supplied in pearl sha pe, melting point 60-62 degrees
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Paraffin waxes and Hydrocarbon waxes