missing translation for 'onlineSavingsMsg'
Learn More
Learn More
His Tag Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 44 publications
Brand: R&D Systems MAB050-100
Additional Details : Weight : 0.00970kg
Description
His Tag Monoclonal specifically detects His Tag in Multi-Species samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunoaffinity Purification, CyTOF-ready.Specifications
His Tag | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Detects proteins containing accessible consecutive histidine regions. The antibody detects His tags localized at the amino- or carboxyl-terminus. | |
Multi-species | |
Supernatant |
Western Blot, Flow Cytometry, Affinity Purification, CyTOF | |
AD1.1.10 | |
Western Blot 0.2 ug/mL, Simple Western 10 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunoaffinity Purification, CyTOF-ready | |
6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, polyHistidine, Poly-histidine | |
His-tagged peptide | |
100 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |